Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™

Supplier: BURDICK & JACKSON FKA SIGMA
Danger

71690-1KG 71690-500G 71690-5KG
BJ71690-1KGEA 188.94 USD InStock
BJ71690-1KG BJ71690-500G BJ71690-5KG
Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™
Sodium hydroxide


Formula: NaOH
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 98.0 %
Assay of Na₂CO₃ Max. 1.0 %
Silver (Ag) Max. 0.0005 %
Aluminium (Al) Max. 0.0005 %
Barium (Ba) Max. 0.0005 %
Bismuth (Bi) Max. 0.0005 %
Calcium (Ca) Max. 0.0005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Mercury (Hg) Max. 0.00001 %
Potassium (K) Max. 0.02 %
Lithium (Li) Max. 0.0005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Molybdenum (Mo) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Strontium (Sr) Max. 0.0005 %
Thallium (Tl) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Ag) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Total N Max. 0.0003 %
Phosphate (PO₄) Max. 0.0005 %
Silicate (as SiO₂) Max. 0.0005 %
Sulfate (SO₄) Max. 0.003 %
Vapor Pressure 3 mmHg ( 37 °C)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty