Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥95%

Supplier: MATRIX SCIENTIFIC MS
Danger

097891-1G 097891-5G
103675-658EA 150.77 USD InStock
103675-658 103675-660
2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥95%
2-[2-(2-Aminoethoxy)ethoxy]ethanol


Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Density: 1.0773 g/cm³
MDL Number: MFCD07367495
CAS Number: 6338-55-2
UN: 2735
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty