Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-chlorophenol-6-sulfonic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

A1448-100GM A1448-25GM
75871-744EA 359.35 USD InStock
75871-744 75871-746
2-Amino-4-chlorophenol-6-sulfonic acid
2-Amino-4-chlorophenol-6-sulfonic acid


Formula: C₆H₆ClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD00035768
CAS Number: 88-23-3
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty