Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butoxyphenol

Supplier: SPECTRUM CHEMICAL MFG CORP

B2159-100GM B2159-25GM
75872-042EA 328.35 USD InStock
75872-042 75872-044
4-Butoxyphenol
4-Butoxyphenol


Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00002336
CAS Number: 122-94-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty