Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Betaine hydrochloride

Supplier: SPECTRUM CHEMICAL MFG CORP

BE175-100GM BE175-1KG BE175-12KGBL
75872-074EA 131.65 USD InStock
75872-074 75872-076 76014-260
Betaine hydrochloride
Betaine hydrochloride
Betaine Hydrochloride is a vitamin-like substance that is found in many whole grains. It is an acidic form of betaine, which is a form of hydrochloric acid that is derived from the beet. It can be used as a supplement to aid in the digestion of proteins as well as other benefits. It is used as a moisturizer and stabilizer in skin care formulations and increases the water solubility of active compounds. In surfactant solutions, betaine acts as a foam modifier and increases the foam volume and stability and improves the after-feel of conditioners.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₅H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00011903
CAS Number: 590-46-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty