Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl salicylate

Supplier: SPECTRUM CHEMICAL MFG CORP

I1342-100ML I1342-25ML
75872-796EA 318.85 USD InStock
75872-796 75872-798
Isopropyl salicylate
Isopropyl salicylate


Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00035703
CAS Number: 607-85-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty