Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl phenylglyoxylate

Supplier: SPECTRUM CHEMICAL MFG CORP

M1595-100GM M1595-25GM
75872-956EA 301.2 USD InStock
75872-956 75872-958
Methyl phenylglyoxylate
Methyl phenylglyoxylate


Formula: C₉H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00008443
CAS Number: 15206-55-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty