Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (±)-lactate

Supplier: SPECTRUM CHEMICAL MFG CORP

M1716-100ML M1716-25ML M1716-500ML
75872-966EA 108.55 USD InStock
75872-966 75872-968 75872-970
Methyl (±)-lactate
Methyl (±)-lactate


Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 35
Storage Temperature: Ambient
MDL Number: MFCD00066367
CAS Number: 547-64-8
UN: 3272
ADR: 3 III
Merck Index: 12,06167
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty