Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-1,3-benzodioxole

Supplier: SPECTRUM CHEMICAL MFG CORP

M2303-100GM M2303-25GM
75873-020EA 445 USD InStock
75873-020 75873-022
5-Nitro-1,3-benzodioxole
5-Nitro-1,3-benzodioxole


Formula: C₇H₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00005824
CAS Number: 2620-44-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty