Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-α-Phenylalanine

Supplier: SPECTRUM CHEMICAL MFG CORP

PH143-100GM PH143-1KGBL PH143-25GM
75873-262EA 402.95 USD InStock
75873-262 75873-264 75873-266
D-α-Phenylalanine
D-α-Phenylalanine


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 295
Storage Temperature: Ambient
MDL Number: MFCD00004270
CAS Number: 673-06-3
Merck Index: 12,07425
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty