Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethoxybenzoic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

V3057-100GM V3057-25GM
75873-548EA 234.75 USD InStock
75873-548 75873-550
3,4-Dimethoxybenzoic acid
3,4-Dimethoxybenzoic acid


Formula: C₉H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00002500
CAS Number: 93-07-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty