Human Recombinant Apo-SAA1 (from E. coli)
Supplier: PEPROTECH INC.
AF-300-53-10UG AF-300-53-50UG AF-300-53-100UG AF-300-53-250UG AF-300-53-500UG AF-300-53-1MG
76303-644EA 158.05 USD InStock
76303-644 76303-646 76304-168 76304-170 76304-172 76304-174
Human Recombinant Apo-SAA1 (from E. coli)
Proteins and Peptides
Serum amyloid A proteins (SAA) represents a family of apolipoproteins that circulates in association with high-density lipoproteins (HDL). The level of Apo-SAA, normally 1-5 ug/ml in plasma, increases 500-1000 fold within 24 hours of an inflammatory stimulus and, under these conditions, is the most abundant HDL apolipoprotein. The human SAA gene codes for a 122 amino acid nonglycosylated polypeptide, which contains an 18 amino acid N-terminal sequence. Recombinant Human Apo-SAA1 is an 11.7 kDa protein containing 105 amino acid residues.
  • Animal-free
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty