Condensers, Cold Finger, Chemglass
Supplier: Chemglass
CG-1222-01 CG-1222-04 CG-1222-05 CG-1222-14
80067-430EA 162.6 USD InStock
80067-430 80067-434 80067-436 80067-438
Condensers, Cold Finger, Chemglass
Condensers
Condenser has an inner joint and a sealed lower drip tip for drop control
Hose connections have an O.D. of 10 mm at the middle serration.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty