Support Rings, with Openings, Chemglass
Supplier: Chemglass
CG-1948-L-11 CG-1948-L-12
10034-378EA 7.53 USD InStock
10034-378 10034-380
Support Rings, with Openings, Chemglass
Support Rings
Support rings with openings.
  • Manufactured from steel wire
  • And zinc plated rings
Support ring have an opening to permit items such as separatory funnels to be inserted without removing the support ring from the base.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty