Reaction Vessels, Cylindrical, Jacketed, Morton Indents, Lower Drain Valves, Chemglass
Supplier: Chemglass
CG-1931-23 CG-1931-24 CG-1931-25 CG-1931-26 CG-1931-27 CG-1931-20 CG-1931-22
47733-850EA 2169.17 USD InStock
47733-850 47733-852 47733-854 47733-856 47733-858 80065-470 80071-526
Reaction Vessels, Cylindrical, Jacketed, Morton Indents, Lower Drain Valves, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Reaction vessels feature morton indentations.
  • Similar to CG-1930, but with morton indentations to increase agitation during stirring
  • Supplied complete with two polypropylene hose connections and two screw caps
For clamp see CG-141 or CG-141-T. For O-ring see CG-147. Morton style reactors/flasks are not suitable for vacuum.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty