Distilling Columns, Water Jacketed, Chemglass
Supplier: Chemglass
CG-1230-05 CG-1230-06 CG-1230-08 CG-1230-09
80067-506EA 166.08 USD InStock
80067-506 80067-508 80067-512 80067-514
Distilling Columns, Water Jacketed, Chemglass
Distillation Components Distillation Columns
Distillation columns feature water jacket.
  • Jacketed distilling column has indentations on the inner tube to support packing material
The jacket allows the column to be used as a condenser. Column has a top outer and lower inner standard taper joint. All sizes have an I.D. of approximately 10 mm.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty