Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Petroleum spirit, 40…60 °C ≥90%, Puriss. p.a. ACS, ISO, low boiling point hydrogen treated naphtha, Riedel-de Haën™

Supplier: BURDICK & JACKSON FKA SIGMA
Danger

32299-195L 32299-1L 32299-2.5L 32299-4X2.5L 32299-4X5L 32299-5L 32299-6X1L
BJ32299-195LEA 3498.61 USD InStock
BJ32299-195L BJ32299-1L BJ32299-2.5L BJ32299-4X2.5L BJ32299-4X5L BJ32299-5L BJ32299-6X1L
Petroleum spirit, 40…60 °C ≥90%, Puriss. p.a. ACS, ISO, low boiling point hydrogen treated naphtha, Riedel-de Haën™
Petroleum spirit
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

RTECS Number: OI6180000
UNSPSC Code: 12191502


Boiling Pt: 40…60 °C (1.013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 309-852-0
UN: 3295
ADR: 3,II
Order Now
Specification Test Results
Boiling range 40-60 °C Min. 90 %
Boiling range 40 - 60 °C
Density (D 20/4) 0.640 - 0.655
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.002 %
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Benzene (GC) Max. 0.01 %
Iodine value Max. 0.3
S-compounds (as S) Max. 0.005 %
APHA Max. 10

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty