Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dioxane ≥99.5% (by GC) stabilized, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: BURDICK & JACKSON FKA SIGMA
Danger

33147-1L 33147-2.5L 33147-210KG
BJ33147-1LEA 252.46 USD InStock
BJ33147-1L BJ33147-2.5L BJ33147-210KG
1,4-Dioxane ≥99.5% (by GC) stabilized, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
1,4-Dioxane
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 102551
RTECS Number: JG8225000
UNSPSC Code: 12191602


Formula: C₄H₈O₂
Boiling Pt: 100-102 °C (1.013 hPa)
Melting Pt: 12 °C
Density: 1.031…1.034 g/cm³ (20 °C)
Flash Pt: 22.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=64c930c52bed4490a4dd38741bf8cb01a966ce0f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) Min. 99.5 %
Congealing - Freezing Point Min. 11 °C
Boiling range 100 - 102 °C
Density (D 20/4) 1.031 - 1.034
Refractive index (n 20/D) 1.4210 - 1.4240
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Acetaldehyde Max. 0.005 %
Carbonyl Compounds (as HCHO) Max. 0.01 %
Peroxides (as H₂O₂) Max. 0.005 %
APHA Max. 10

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty