Oakton® Conductivity Calibration Pouches, Cole Parmer
Supplier: ***DO NOT USE***
35653-08 35653-09 35653-10 35653-13 35653-14
76338-522PK 68.16 USD InStock
76338-522 76338-524 76338-526 76338-530 76338-532
Oakton® Conductivity Calibration Pouches, Cole Parmer
Standards Conductivity Standards
Conductivity pouches sealed for freshness and easy travel.
  • Single use pouches
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
Single-use 20-mL pouches are sealed to ensure freshness—take these convenient standard solutions anywhere. Don't worry about transferring solution into another container—calibrate directly from the pouch.
A µS-to-ppm conversion table is printed on each pack for quick reference.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty