Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl sulfoxide ≥99.9%, Cryopres™ USP for cryopreservation, sterile-filtered

Supplier: E. Becker & Co. GmbH
Warning

092780145 092780148
76337-180EA 57.58 USD InStock
76337-180 76337-182
Dimethyl sulfoxide ≥99.9%, Cryopres™ USP for cryopreservation, sterile-filtered
Dimethyl sulphoxide
Cryopres™ Dimethyl sulfoxide is commonly used in cell cryopreservation, along with glycerol, to prevent damage to cell membranes during freezing. USP, Ph. Eur., sterile-filtered.

Precaución: For laboratory use. Not for injection.


Formula: (CH₃)₂SO
Boiling Pt: 189 °C (lit.)
Melting Pt: 16…19 °C (lit.)
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 87 °C (188.6 °F) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty