Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD172a (SIRPalpha) Rat Monoclonal Antibody [clone: P84]

Supplier: Bio X Cell
BE0322-1MG BE0322-25MG BE0322-5MG BE0322-50MG BE0322-100MG
103778-438EA 3920 USD InStock
103778-438 103778-440 103778-442 103778-444 103778-446 76554-688 76556-182 76556-494 76556-508 76557-016
Anti-CD172a (SIRPalpha) Rat Monoclonal Antibody [clone: P84]
Antibodies
Type: Primary
Antigen: CD172a (SIRPalpha)
Clonality: Monoclonal
Clone: P84
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty