Limited Flammability Jackets, MCR Safety
Supplier: MCR Safety
39030S 39030M 39030L 39030XL 39030XXL 39030XXX 39030X4 39030X5
76340-846EA 55.71 USD InStock
76340-846 76340-856 76340-884 76340-898 76340-916 76340-928 76340-950 76340-962
Limited Flammability Jackets, MCR Safety
Workwear
All limited flammability cotton apparel is made of 9 oz. green cotton. The 100% material cotton is cool, comfortable and launderable. These products provide economical protection from light molten splash and sparks.
  • USA treated cotton fabric
  • Anodized button snap closures
These launderable garments maintain limited flammability up to 50 home washings or 25 commercial washings (up to 165 °F). They are ideally suited for use in aircraft manufacturing, construction, demolition, metal fabrication, Mig/Tig, ship building, welding, and general manufacturing applications.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty