Paper Labels, Blank, 4×6" Rectangular, Labelmaster
Supplier: Label Master
BH10C BH10O BH10P BH10G BH10Y
101376-722PK 137.31 USD InStock
101376-722 101376-726 101376-728 101376-724 101376-736
Paper Labels, Blank, 4×6" Rectangular, Labelmaster
Labels
The labeling possibilities are virtually endless with Blank 4×6" Rectangular Paper labels.
  • Thermal transfer imprintable
  • Outstanding chemical and abrasion resistance
  • Strong adhesion to corrugated boxes
Blank labels may be used for countless applications, from identifying inventory and property to marking packages with special handling information.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty