Radioactive III Labels, Labelmaster
Supplier: Label Master
HMSL16 HML16
101377-916PK 130.25 USD InStock
101377-916 101376-368
Radioactive III Labels, Labelmaster
Labels
Employ a fool-proof shipping label solution with Worded Radioactive III labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives are formulated for use in various climates and environments
  • Strong holds on corrugated boxes
  • Once affixed, labels won't fade or fall off
  • Outstanding chemical and abrasion resistance
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty