Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycerine ACS

Supplier: RICCA CHEMICAL CO
Warning

RSOG0020-1A RSOG0020-20F RSOG0020-4A RSOG0020-500A
RCRSOG0020-1AEA 87 USD InStock
RCRSOG0020-1A RCRSOG0020-20F RCRSOG0020-4A RCRSOG0020500A
Glycerine ACS
Glycerine


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 (dec.)
Melting Pt: 18.6
Density: 1.2636
Flash Pt: 199°C
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=481dd90ecbdec73cd4020432d3356161ebfe7681&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay min 99.5 %
Color (APHA) max 10
Neutrality To Pass Test
Acrolein and Glucose To Pass Test
Chlorinated compounds (as Cl) max 0.003 %
Heavy Metals (as Pb) max 2 ppm
Sulfate (SO₄) max 0.001 %
Water (H₂O) max 0.5 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty