AlphaTec® 68-4000 Chemical Barrier Coveralls, Ansell
Supplier: Ansell Healthcare
GR40-T-92-122-05 GR40-T-92-122-06 GR40-T-92-122-07 GR40-T-92-122-08 GR40-T-92-122-09 GR40-T-92-122-04 GR40-T-92-122-02 GR40-T-92-122-03
76020-690CS 616.83 USD InStock
76020-690 76020-692 76020-694 76020-696 76020-698 76045-186 76045-302 76045-304
AlphaTec® 68-4000 Chemical Barrier Coveralls, Ansell
Disposable Garments
Innovative multi-layer chemical barrier technology offering Type 3/4/5 protection.
  • Over 200 chemicals permeations tested, including chemical warfare agents
  • Innovative protective features include dual zip systems and double cuffs
  • Welded and taped seams, offering the highest barrier to liquids and particulates
  • Enhanced comfort; knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static
Engineered to provide an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
Certificaciones: AATCC76; ASTM F1671. CE Category III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty