Heating Blankets, 110 V, Durex Industries
Supplier: Chemglass
CLS-1380-19V CLS-1383-07V CLS-1385-12V
76333-444EA 697.99 USD InStock
76333-444 76333-456 76333-458
Heating Blankets, 110 V, Durex Industries
Heaters Heating Mantles
Manufactured to fit snugly around unjacketed vessels to supply uniform heat.
Made of silicone rubber, all sizes are 110VAC with the wattage as listed. Velcro(-style) allows easy installation, even with the vessel in a support stand. Plugs are IEC standard 2-prong.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty