Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tenofovir, hydrate ≥98% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

T3006-1G T3006-5G
TCT3006-1GEA 55 USD InStock
TCT3006-1G TCT3006-5G
Tenofovir, hydrate ≥98% (by HPLC, titration analysis)
Tenofovir


Formula: C₉H₁₄N₅O₄P
MW: 287.22 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00943794
CAS Number: 147127-20-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd128d963daa7df2acffdba83efa5c0c396231ff&vid=bd128d963daa7df2acffdba83efa5c0c396231ff&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystaline
Purity(HPLC) min. 98.0 area%
Purity(Neutralization titration) min. 98.0 %(calcd.on anh.substance)
Specific rotation [a]20/D -19.0 to -21.0 deg(C=1, 0.1mol/L HCl)(calcd.on anh.substance)
Water max. 6.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty