Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Adonitol

Supplier: Bioanalytic

A32000-100.0 A32000-1000.0 A32000-2000.0 A32000-25.0 A32000-5.0 A32000-500.0
76344-886EA 412.86 USD InStock
76344-886 76344-888 76344-890 76344-892 76344-894 76344-896
Adonitol
Adonitol
Adonitol is a crystalline pentose alcohol formed by the reduction of ribose.

  • It occurs naturally in the plant Adonis vernalis, as well as in the cell walls of Gram positive bacteria.


Formula: HOH₂C(CH(OH))₃CH₂OH
MW: 152.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064291
CAS Number: 488-81-3
EINECS: 207-685-7
Merck Index: 13,00168
Order Now
Specification Test Results
Appearance White to off white powder
Identity (¹H NMR, D20) Conforms to structure
Solubility (100mg/2ml H2O) Colorless to faint yellow

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty