Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium carbonate

Supplier: Bioanalytic

C30020-1000.0 C30020-3000.0 C30020-500.0 C30020-5000.0
76345-364EA 85.85 USD InStock
76345-364 76345-366 76345-368 77510-202
Calcium carbonate
Calcium carbonate


Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658
Order Now
Specification Test Results
Appearance White powder
Chloride ≤ 330 ppm
Loss on Drying ≤ 2.0 %
Magnesium and alkali metals ≤ 1 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty