High Performance FR Short Sleeve Knit Crew, Gray, Lakeland Industries
Supplier: Lakeland
SSCAT06-2X SSCAT06-2XT SSCAT06-3X SSCAT06-3XT SSCAT06-4X SSCAT06-4XT SSCAT06-5X SSCAT06-LG SSCAT06-LGT SSCAT06-MD SSCAT06-MDT SSCAT06-SM SSCAT06-XL SSCAT06-XLT
76439-180EA 125.65 USD InStock
76439-180 76439-182 76439-184 76439-186 76439-188 76439-190 76439-192 76439-194 76439-196 76439-198 76439-200 76439-202 76439-204 76439-206
High Performance FR Short Sleeve Knit Crew, Gray, Lakeland Industries
Workwear
Lightweight, inherently FR Short Sleeve Knit Crew with permanent moisture wicking properties make this an extremely comfortable FR/AR base layer garment.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag-free collar
  • Drop shoulder design
  • Left chest pocket
Base layers are the foundational key to the Lakeland FR/AR Layering System. To keep cool during hot summer days, and warm when it's frigid cold outside, wearers need to be dry. Base layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers to constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certificaciones: NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty