Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-DNP HEK293 Polyclonal Antibody(UltraLys)

Supplier: ACROBIOSYSTEMS INC MS
DNPBM2200UG DNPBM225UG
103870-884EA 1055.38 USD InStock
103870-884 103870-886
Anti-DNP HEK293 Polyclonal Antibody(UltraLys)
Antibodies
Type: Primary
Antigen: DNP
Clonality: Polyclonal
Clone:
Conjugation: UltraLys
Epitope:
Host: HEK293
Isotype: Human IgG1/kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty