Thermolabile Exonuclease I
Supplier: W.u.G. Dickgießer GmbH
M0568L M0568S
76326-032EA 361 USD InStock
76326-032 76326-034
Thermolabile Exonuclease I
Enzymes
Thermolabile Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA (ssDNA) in the 3' to 5' direction.
  • DNA specific exonuclease
  • Unlike Exonuclease I , Thermolabile Exonuclease I can be heat inactivated at 80C in 1 minute
  • Removal of single-stranded primers in PCR reactions prior to DNA sequencing or SNP analysis
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty