Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate 95%

Supplier: AMBEED, INC
Warning

A159435-5G A159435-100G A159435-25G A159435-5G A159435-1000G A159435-500G
103843-474EA 5 USD InStock
103843-474 77028-584 77028-586 77300-402 77319-358 77671-256 77671-258
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate 95%
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate
Precaución: for Research Use Only


Formula: C₁₉H₁₄F₂O₆
Boiling Pt: 437.2 °C (760 mm Hg)
Melting Pt: 117…119 °C
Storage Temperature: Freezer
MDL Number: MFCD08458308
CAS Number: 122111-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty