Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-Sparteine ≥97%

Supplier: AMBEED, INC
Warning

A135377-100G A135377-5G
103802-144EA 21 USD InStock
103802-144 77881-067 77881-068
(+)-Sparteine ≥97%
(+)-Sparteine
Precaución: Keep in dark place, sealed in dry, room temperature


Formula: C₁₅H₂₆N₂
Boiling Pt: 340.9 °C (760 mm Hg)
Flash Pt: 148.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00869353
CAS Number: 492-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty