Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Tetrahydro-2h-pyran-2-yl)-5-(trifluoromethyl)-1h-indazol-4-ylboronic acid 97%

Supplier: AMBEED, INC
Warning

A255679-1G A255679-1G A255679-100MG A255679-250MG
103812-204EA 329 USD InStock
103812-204 77091-522 77250-018 77250-020
1-(Tetrahydro-2h-pyran-2-yl)-5-(trifluoromethyl)-1h-indazol-4-ylboronic acid 97%
1-(Tetrahydro-2h-pyran-2-yl)-5-(trifluoromethyl)-1h-indazol-4-ylboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₄BF₃N₂O₃
MW: 314.068 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30609549
CAS Number: 2374153-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty