Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%

Supplier: AMBEED, INC
Warning

A254450-1G A254450-5G A254450-100MG A254450-250MG A254450-1G A254450-5G
103812-140EA 290 USD InStock
103812-140 103851-994 76692-588 76692-590 77091-436 77091-438
Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%
Chloro(1,5-cyclooctadiene)rhodium (I) dimer
Precaución: for Research Use Only


Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00012415
CAS Number: 12092-47-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty