Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-1,2,4-triazol-5-yl)methanol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A190957-1G A190957-100MG A190957-250MG
103816-338EA 567 USD InStock
103816-338 77151-874 77151-876
(1H-1,2,4-triazol-5-yl)methanol hydrochloride 95%
(1H-1,2,4-triazol-5-yl)methanol hydrochloride
Precaución: for Research Use Only


Formula: C₃H₆ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD29037427
CAS Number: 1195596-30-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty