Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-imidazole-2-boronic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A207301-1G A207301-5G A207301-1G A207301-100MG A207301-250MG
103817-780EA 167 USD InStock
103817-780 103848-460 77303-028 77671-830 77881-487
1H-imidazole-2-boronic acid hydrochloride 95%
1H-imidazole-2-boronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₃H₅BN₂O₂·ClH
Storage Temperature: Freezer
MDL Number: MFCD16251134
CAS Number: 1919022-57-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty