Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Methyl-1H-pyrazol-3-yl)methanol 98%

Supplier: AMBEED, INC
Warning

A130924-1G A130924-1G A130924-5G
103801-494EA 20 USD InStock
103801-494 77313-972 77313-974
(1-Methyl-1H-pyrazol-3-yl)methanol 98%
(1-Methyl-1H-pyrazol-3-yl)methanol
Precaución: for Research Use Only


Formula: C₅H₈N₂O
MW: 112.13 g/mol
Boiling Pt: 243.6 °C (760 mm Hg)
Flash Pt: 101.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08690274
CAS Number: 84547-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty