Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Methyl-1H-pyrazol-4-yl)methanol 98%

Supplier: AMBEED, INC
Warning

A317930-1G A317930-5G A317930-100G A317930-10G A317930-25G
103825-154EA 15 USD InStock
103825-154 103836-084 76838-930 76838-932 77114-690 77114-692 77114-694
(1-Methyl-1H-pyrazol-4-yl)methanol 98%
(1-Methyl-1H-pyrazol-4-yl)methanol
Precaución: for Research Use Only


Formula: C₅H₈N₂O
Boiling Pt: 243 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01822311
CAS Number: 112029-98-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty