Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid 97%

Supplier: AMBEED, INC
Warning

A153990-1G A153990-5G A153990-100MG A153990-10G A153990-1G A153990-5G A153990-250MG
103804-590EA 36 USD InStock
103804-590 103842-770 76871-762 76871-764 77318-454 77318-456 77671-164
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid 97%
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid
Precaución: for Research Use Only


Formula: C₅H₆BF₃N₂O₂
Boiling Pt: 311.3 °C (760 mm Hg)
Melting Pt: 132…137 °C
Flash Pt: 142.04 °C
Storage Temperature: Freezer
MDL Number: MFCD08701785
CAS Number: 344591-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty