Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2R)-2-Aminocyclohexanol 98%

Supplier: AMBEED, INC
Danger

A511972-1G A511972-10G A511972-5G A511972-1G A511972-100G
103828-404EA 13 USD InStock
103828-404 103839-504 76790-022 76843-492 77307-660 77669-274
(1R,2R)-2-Aminocyclohexanol 98%
(1R,2R)-2-Aminocyclohexanol
Precaución: for Research Use Only


Formula: C₆H₁₃NO
Boiling Pt: 201.1 °C (760 mm Hg)
Flash Pt: 75.434 °C
Storage Temperature: Ambient
MDL Number: MFCD08061325
CAS Number: 931-16-8
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty