Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2S)-2-Fluorocyclopropanamine-4-methylbenzenesulfonate 97%

Supplier: AMBEED, INC
Warning

A105983-1G A105983-5G A105983-25G A105983-1G A105983-5G
103796-982EA 11 USD InStock
103796-982 103822-732 76821-554 77309-594 77309-596
(1R,2S)-2-Fluorocyclopropanamine-4-methylbenzenesulfonate 97%
(1R,2S)-2-Fluorocyclopropanamine-4-methylbenzenesulfonate
Precaución: for Research Use Only


Formula: C₁₀H₁₄FNO₃S
Boiling Pt: 418.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01861147
CAS Number: 143062-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty