Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2S)-trans-2-Aminocyclohexanol hydrochloride 97%

Supplier: AMBEED, INC
Warning

A165056-1G A165056-5G A165056-100G A165056-10G A165056-1G A165056-25G A165056-5G
103805-940EA 6 USD InStock
103805-940 103844-198 76973-752 76973-754 77300-760 77320-392 77320-394
(1S,2S)-trans-2-Aminocyclohexanol hydrochloride 97%
(1S,2S)-trans-2-Aminocyclohexanol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO
Boiling Pt: 201.1 °C (760 mm Hg)
Flash Pt: 75.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09259963
CAS Number: 13374-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty