Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Hydroxymethyl)-2-(methylthio)pyrimidine 97%

Supplier: AMBEED, INC
Warning

A185152-1G A185152-5G A185152-250MG A185152-1G
103815-826EA 83 USD InStock
103815-826 103846-394 76825-062 77301-956
5-(Hydroxymethyl)-2-(methylthio)pyrimidine 97%
5-(Hydroxymethyl)-2-(methylthio)pyrimidine
Precaución: for Research Use Only


Formula: C₆H₈N₂OS
MW: 156.206 g/mol
Boiling Pt: 322.8±17.0 °C (760 mm Hg)
Melting Pt: 59…61 °C
Flash Pt: 149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11053756
CAS Number: 19858-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty