Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethoxybenzylamine 98%

Supplier: AMBEED, INC
Danger

A273191-5G A273191-10G A273191-25G A273191-5G A273191-100G
103853-052EA 9 USD InStock
103853-052 76816-252 77092-848 77092-850 77672-412
2,4-Dimethoxybenzylamine 98%
2,4-Dimethoxybenzylamine
Precaución: for Research Use Only


Formula: C₉H₁₃NO₂
Boiling Pt: 271 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052393
CAS Number: 20781-20-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty