Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethoxybenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A137890-100G A137890-10G A137890-500G A137890-25G
77246-770EA 69 USD InStock
77246-770 77246-772 77246-774 77315-512
2,4-Dimethoxybenzyl alcohol 98%
2,4-Dimethoxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₉H₁₂O₃
Boiling Pt: 316.7 °C (760 mm Hg)
Melting Pt: 35…39 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004614
CAS Number: 7314-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty