Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichlorophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A175557-5G A175557-25G A175557-100G A175557-10G A175557-1G A175557-5G
103845-366EA 9 USD InStock
103845-366 77082-710 77273-856 77273-858 77273-860 77301-380
2,6-Dichlorophenylboronic acid 97%
2,6-Dichlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BCl₂O₂
Boiling Pt: 341.8 °C (760 mm Hg)
Melting Pt: 153 °C
Flash Pt: 160.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00064869
CAS Number: 73852-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty