Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorophenylhydrazine 97%

Supplier: AMBEED, INC
Warning

A222173-1G A222173-5G A222173-1G A222173-5G A222173-250MG
103818-968EA 59 USD InStock
103818-968 103849-844 77088-446 77088-448 77288-242
2,6-Difluorophenylhydrazine 97%
2,6-Difluorophenylhydrazine
Precaución: for Research Use Only


Formula: C₆H₆F₂N₂
Boiling Pt: 168.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06658266
CAS Number: 119452-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty